Learn More
Sartorius Qualitative Grade 5 H/N Filter Papers
This paper grade is generally used for analytical and technical purposes. It has a fast flow rate due to its structure.
Brand:  Sartorius FT-4-423-150
Additional Details : Weight : 0.10000kg
Product Code. 10545701
  • 129.00€ / Pack of 100
Estimated Shipment: 29-05-2024
to see stock.
Add to basket
Description
Description
  • Made of refined pulp and cotton linters with an > 95% α-cellulose content
  • Ash content between 0.1 - 0.15%
  • Wet-strengthened
Specifications
Specifications
Folded
5 H/N
0.28 mm
3 sec.
100/Pk.
150 mm