Description
arium™ refill pack of ion exchange resin mix in 25l PE bag for complete deionization cartridges
Specifications
Specifications
| For Use With (Equipment) | 615DI Deionization Cartridges |
| Type | Ion Exchange Resin |
| For Use With (Application) | Remove cations and anions from the water by exchanging H+ and OH- |
Product Title
Haben Sie Verbesserungsvorschläge?