Learn More
Sartorius Cubis™ Balance Display and Control Units
Shop All Sartorius Products
Click to view available options
Includes:
Backlit black and white graphic display and tactil
Color TFT graphic display and touch screen
Unit Size:
Each
Description
For use with Cubis™ balances.
Specifications
Specifications
| For Use With (Equipment) | Cubis Balance |
| Includes | Backlit black and white graphic display and tactil |
| Type | Control Unit |