Learn More
Sartorius Minisart™ NML Syringe Filters, Sterile
Product Code. p-4901723 Shop All Sartorius Products
Change view
Click to view available options
Pore Size:
0.2 μm
0.45 μm
0.65 μm
0.8 μm
1.2 μm
5 μm
Color:
Blue
Brown
Green
Pink
Red
Yellow
Outlet Style:
Male Luer Lock
Male Luer Slip
Unit Size:
Pack of 50
Sterilization Method:
ETO-sterile
Gamma-sterile, individually packaged
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size Color Outlet Style Unit Size Sterilization Method
11795745 0.2 μm Pink Male Luer Lock Pack of 50 -
10509821 0.2 μm Blue Male Luer Lock Pack of 50 ETO-sterile
10076891 0.2 μm Blue Male Luer Lock Pack of 50 Gamma-sterile, individually packaged
10527401 0.2 μm Blue Male Luer Slip Pack of 50 ETO-sterile
11736035 0.65 μm Pink Male Luer Lock Pack of 50 ETO-sterile
10009230 0.45 μm Yellow Male Luer Slip Pack of 50 ETO-sterile
11786953 0.45 μm Yellow Male Luer Lock Pack of 50 Gamma-sterile, individually packaged
10109180 0.45 μm Yellow Male Luer Lock Pack of 50 ETO-sterile
10413131 0.8 μm Green Male Luer Lock Pack of 50 ETO-sterile
11746035 0.8 μm Green Male Luer Lock Pack of 50 Gamma-sterile, individually packaged
10189220 1.2 μm Red Male Luer Lock Pack of 50 ETO-sterile
10230891 5 μm Brown Male Luer Lock Pack of 50 ETO-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 11795745 Supplier Sartorius Supplier No. 17528K
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Provides fast and reliable filtration of small to medium volumes, complete with ready-to-connect filter units. Sartorius Minisart™ NML Syringe Filters, Sterile eliminate time consuming preparatory work, such as changing out the filter and cleaning the filtration system.
Fast and reliable filtration of small to medium volumes, complete with ready-to-connect filter units eliminate time consuming preparatory work, such as changing out the filter and cleaning the filtration system. These labor saving units are used for a wide variety of applications: sample cleaning, particle removal, and sterile filtration.
  • Handy and readily available
  • Easy to use
  • Proven quality
  • Economical
  • Available in a wide array of different types
  • 100% tested during manufacture
TRUSTED_SUSTAINABILITY
Specifications
Color Pink
Disposable Single-use
Filter Type Syringe Filter
Material (Membrane) SFCA
Pore Size 0.2 μm
Sterility Sterile
Filtration Area 6.2 cm2
For Use With (Application) Preparation of aqueous liquids
Hold up Volume (After Air Purge) 0.15 mL
Housing Material Meta acrylate butadiene styrene polymerisate (MBS)
Inlet Style Female Luer Lock
Outlet Style Male Luer Lock
Packaging ETO-sterile
Prefilter No
Quantity 50 Pk.
Temperature (Metric) Max 50°C
Type Ophthalsart Syringe Filter
Volume (Metric) 200 mL
Diameter (Metric) 28 mm
For Use With (Equipment) Syringe
Sample Volume 1 to 200 mL