Learn More
Sartorius Quantitative Grade 1289 Filter Papers
These qualitative filter papers are essentially used for analytical purposes and routine analyses, whenever no gravimetric analyses are required. They are wet-strengthened and can be used for pressure and vacuum filtration.
Brand:  Sartorius FT-3-207-125
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10480272
  • 20.70€ / Pack of 100
In Stock
to see stock.
Add to basket
Description
Description
Made of refined pulp and cotton linters with an > 95% α-cellulose content. Ash content < 0,1% according to DIN 54370. Wet-strengthened.
Specifications
Specifications
Disc
20 sec.
100/Pk.
125 mm
8 to 12 μm
1289
0.21 mm