Learn More
Sartorius Quantitative Grade 292 Filter Papers
These filter papers are especially suitable for qualitative analyses that require a low ash content.
Brand:  Sartorius FT-3-205-090
Additional Details : Weight : 0.06392kg
 View more versions of this product
Product Code. 10594751
  • 12.30€ / Pack of 100
Estimated Shipment: 18-06-2024
to see stock.
Add to basket
Description
Description
  • Pure cotton linters or cotton linters with refined pulp
  • No additives, such as wet-strengthening agents
Specifications
Specifications
Disc
60 sec.
100/Pk.
90 mm
5 to 8 μm
292
0.18 mm