Learn More
Sartorius Quantitative Grade 292 Filter Papers
These filter papers are especially suitable for qualitative analyses that require a low ash content.
Brand:  Sartorius FT-3-205-030
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10721563
  • 9.00€ / Pack of 100
Estimated Shipment: 05-07-2024
to see stock.
Add to basket
Description
Description
  • Pure cotton linters or cotton linters with refined pulp
  • No additives, such as wet-strengthening agents
Specifications
Specifications
Disc
60 sec.
100/Pk.
30 mm
5 to 8 μm
292
0.18 mm