Learn More
Thermo Scientific™ Savant™ DNA SpeedVac™ Concentrator Kits
Save time and money with Thermo Scientific™ Savant™ DNA SpeedVac™ Concentrators Kits. Everything you need for quick drying and concentrating of small sample volumes.
Brand: Thermo Scientific™ DNA120-230
Additional Details : Weight : 37.00000kg
Description
Save time and money with compact Thermo Scientific™ Savant™ DNA SpeedVac™ Concentrators Kits. These complete systems provide everything you need to quickly dry and concentrate small sample volumes in one fully integrated package, sold under one part number.
Operating Features
- Consistent performance: designed to ensure consistent drying times and reproducible results
- Simplicity: single program supports all DNA/RNA applications
- User-friendly interface: with temperature/time settings
- Three drying rates: ambient, ∽43°C, ∽65°C
- Dual digital timers: independent control of heat/run times
Sample Protection
- Dry at ambient temperatures or at very low heat to avoid sample denaturing
- Oil-free diaphragm pump provides gentle transition from atmosphere to ultimate vacuum setting with no bumping
Safety Features
- PTFE-coated chamber resists chemicals and cleans easily
- Latched lid ensures user safety during operation
- Unit and pump are virtually maintenance-free
- Optional chemical traps and cartridges trap radioactivity and ammonia
Solvents and Solvent Combinations:
DNA120- PCR buffers (aqueous)
- Water
- Ethanol and water
- Ammonium hydroxide
- Isopropanel
- PCR buffers (aqueous)
ISS-110
- PCR buffers (aqueous)
- Ethanol and water
- Isopropanol
DNA 120 SpeedVac Kit
- Corrosion-resistant, PTFE coated chamber provides years of lasting service
- Built-in, oil free vacuum pump provides maintenance-free operation
- Integrated and compact design
- Extremely quiet operation
- Choice of three drying rates
- Dual digital timers for independent control of heat and run times
DNA120OP SpeedVac Kit for oligos
- All the features of the DNA 120
- Plus external post trap to neutralize ammonium hydroxide, chemically resistant glass cover to contain solvents
High capacity DNA SpeedVac Kit ISS110
- Built-in ETFE coated vacuum pump providing years of lasting service
- Four liters, -50°C refrigerated vapor trap (integrated)
- Choice of three drying rates
Includes:
DNA120
- DNA concentrator with integrated pump
- RD36 rotor for 36 × 1.6mL microtubes
- DNA concentrator with integrated pump and glass cover
- RD36 rotor for 36 × 1.5mL microtubes
- ANT100 chemical trap
- ANS121 ammonium neutralizing solution
Specifications
CE (230V only) | |
50 Hz | |
230 V | |
-55°C | |
DNA120 |
31 L/min. at 50 Hz, 36 L/min. at 60 Hz | |
7 torr (9 mbar) | |
4 L | |
PTFE | |
230 V 50 Hz |