Learn More
Thermo Scientific™ Savant™ SpeedVac™ Rotors for Large Capacity Concentrators
Shop All Thermo Scientific Products
Click to view available options
Holds:
1.5 mL Microtube, 12 x 75 mm, 13 x 100 mm Glass and Plastic Tubes
12 Deep-well Plates
192 Vials
30 x 15 mL Conical Centrifuge Tubes
52 x 15 mL Conical
8 Microwell Plates
Flasks
Unit Size:
Each
Pack of 2
Description
SpeedVac Rotors for Large Capacity Concentrators for use with Savant SpeedVac Concentrators.
Circular Rotors:
- Easy to use
- Multi-size tube holes accommodate different sample sizes
- Require no additional accessories
- Fit directly into your concentrator
Specifications
Specifications
| Description | Flasks: 2 x 100mL round bottom, Large capacity models |
| Type | Rotor |
| For Use With (Equipment) | SPD210, SPD2030, SPD300DDA (requires RAD200) |
| Holds | Flasks |
| Model | RBA2-FLSK-100 |
| Rotor System | Traditional |