Learn More
Thermo Scientific™ Schaedler Broth (with Haemin and Vitamin K) (Tube)
Description
Cultivate fastidious aerobic and anaerobic microorganisms with Thermo Scientific™ Oxoid™ Schaedler Broth with Haemin and Vitamin K ready prepared medium tubes. Schaedler Broth with Haemin and Vitamin K tubes can also be used to determine antibiotic minimum inhibitory concentration (MIC) levels of anaerobic organisms. The presence of Haemin and vitamin K in the medium supply growth factors required by many fastidious organisms.
Remove the need for autoclaving and dispensing with convenient, pre-filled and ready prepared tubes of Schaedler Broth with Haemin and Vitamin K.
- Enhanced growth of demanding anaerobic organisms compared to seven other commonly used broth media.1
- Convenient: the pre-filled tubes are ready for immediate use.
- Simple: easy-to-use with minimum training.
- Long shelf life: up to 12 months
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
| Color | Golden yellow, transparent |
| Content And Storage | 2°C to 12°C |
| Description | Schaedler Broth (with Haemin and Vitamin K) (Tube) |
| Format | Poured tube |
| Manufacturing Origin | Germany |
| Product Type | Microbiology Media |
| Packaging Type | 50 tubes in a box |
| Quantity | 50/Pk. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.