Learn More
Kartell™ PE Standardized Screw Cap
missing translation for 'orderingAttributeHoverText'
Closure Size:
GL25
Description
- Ligtweight
- For use with wash bottles
- For Kartell™ Bottles 15801. 15821, 15841, 15861, 15881, 15901
Specifications
Material | Polyethylene |
Type | Standardized Screw Cap |
Product Line | Cap |
Closure Size | GL25 |
Closure Material | Polyethylene |
For Use With (Equipment) | Kartell™ Bottles 15801, 15821, 15841, 15861, 15881, 15901 |