Learn More
JG Finneran Associates™ Screw Caps
Click to view available options
Color:
Black
Red
Royal Blue
Material:
PTFE, Rubber
PTFE, Silicone
Closure Size:
13-425
8-425
9 mm
Unit Size:
Pack of 100
Pack of 1000
Description
- The patented Top Hat™ septa and closure provide a positive physical lock to assure that preassembled components arrive intact
- Preassembled R.A.M.™ Smooth Screw Thread Closure and Septa
- Designed to work in Agilent or other robotic arm autosamplers
- Available in a variety of colors
Specifications
Specifications
| Diameter (Metric) Cap | 9 mm |
| Color | Royal Blue |
| Material | PTFE, Rubber |
| Septum | PTFE/Butyl Rubber |
| Quantity | 1000 Pk. |
| Type | Screw Cap |
| Closure Size | 9 mm |
| For Use With (Equipment) | Agilent or Other Robotic Arm Autosamplers |