missing translation for 'onlineSavingsMsg'
Learn More

JG Finneran Associates™ Screw Caps

Product Code. 12423020
Click to view available options
Color:
Black
Red
Royal Blue
Material:
PTFE, Rubber
PTFE, Silicone
Closure Size:
13-425
8-425
9 mm
Unit Size:
Pack of 100
Pack of 1000
This item is not returnable. View return policy

Product Code. 12423020

Brand: JG Finneran Associates™ 5396F09RB

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Use these preassembled caps for a variety of applications. JG Finneran™ JG Finneran™ Screw Caps provide a positive physical lock to assure that preassembled components arrive intact.

  • The patented Top Hat™ septa and closure provide a positive physical lock to assure that preassembled components arrive intact
  • Preassembled R.A.M.™ Smooth Screw Thread Closure and Septa
  • Designed to work in Agilent or other robotic arm autosamplers
  • Available in a variety of colors

Specifications

Diameter (Metric) Cap 9 mm
Color Royal Blue
Material PTFE, Silicone
Septum PTFE/Silicone
Quantity 1000 Pk.
Type Screw Cap
Closure Size 9 mm
For Use With (Equipment) Agilent or Other Robotic Arm Autosamplers
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.