missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Screw Caps For Wheaton™ E-Z Vials™

For 12mm x 32mm E-Z Vials™

Brand:  DWK Life Sciences 242765

Additional Details : Weight : 0.63500kg

 View more versions of this product

Product Code. 12645010

  • 174.00€ / Pack of 1000
Estimated Shipment: 27-05-2024
to see stock.
Add to basket

Description

Description

  • Screw Cap
  • Polypropylene
  • 10mm (0.39in.)
  • Unlined
  • With solid top
  • Black

Specifications

Specifications

Linerless
Polypropylene
10-425
Black
Without top seal
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions