Learn More
OHAUS™ Sectional Block
Shop All Ohaus Products
Click to view available options
For Use With (Equipment):
12 mm Test Tubes
12 mm Vials
15 mm Vials
16 mm Test Tubes
17 mm Vials
20 mm Test Tubes
21 mm Vials
25 mm Test Tubes
28 mm Vials
Unit Size:
Each
Description
Guardian accessories allow end users to have superb versatility. They can create Mini Lab Frames utalizing our large selection of accessories such as Rods, Clamps, Lab Lifts, Uni Blocks, Sectional Blocks, Splash Shields, Stir Bars, Temperature Probes, Achiever Overhead Stirrers
Specifications
Specifications
| For Use With (Equipment) | 12 mm Test Tubes |
| Type | Sectional Block |
| Material | Metal |