Learn More
ITS Thorsten Betzel™ Semiquantitative Strips
Product Code. p-8000281
Click to view available options
Analytes:
Chloride
Free Chlorine
Hydrogen Peroxide
Nitrate-Nitrite/NO3
Total Chlorine
Concentration:
0-0.05-0.1-0.15-0.2-0.5-0.8-1.0-4.0-10.0 ppm
0-0.05-0.1-0.2-0.4-0.6-0. 8-1.2-1.5-2.0-2.6-4.0-> 6.0 ppm
0-5-10-20-30-40-60-80 ppm
0-50-100-250-500 ppm
0. 0.5. 2. 5. 10. 20. 50 ppm(mg/L)
0. 1.0. 2.0. 3.0. 4.0. 5.0. 6.0. 7.0 ppm(mg/L) and 0.00. 0.05. 0.10. 0.20. 0.40. 0.60. 0.80. 1.20 ppm(mg/L)
0.05-0.3-0.5-1.0-2.0-4.0 ppm
Unit Size:
Pack of 50
Product Code. 11752957
Brand: ITS Thorsten Betzel™ 480009
Only null left
Specifications
Analytes Nitrate-Nitrite/NO3
Concentration 0. 0.5. 2. 5. 10. 20. 50 ppm(mg/L)
Packaging Bottle
Quantity 50 Strips