missing translation for 'onlineSavingsMsg'
Learn More

SemperIt™ Semperguard™ Sempercare Skin2 Nitrile Examination Gloves

Wearing comfort and skin-friendliness, a good tactile feeling is the most important assessment criterion when choosing the right exam glove. The new Sempercare Nitrile skin2 combines these properties in a unique, exceptionally light glove.

Brand:  SemperIt™ 3000002157

Additional Details : Weight : 0.01000kg

Product Code. 15303834

  • 50.90€ / Pack of 200
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Nonsterile, powder-free examination glove for single use
  • Nitrile-Butadiene-Rubber (NBR)
  • Lavender
  • Flat with rolrand
  • EN 455-2
  • According to EN 455-1: AQL 1.5
  • According to EN 455-2:
    • Tensile strength Production value ≥ 6 N
    • Median 7 N
  • EN 1041, EN 980, EN 420 and ISO 15223
  • Medical product class I
  • Protective glove for single use
  • PPE category III
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions