missing translation for 'onlineSavingsMsg'
Learn More

Semperit™ Semperguard™ Style Nitrile Gloves

Catalog Number 15593536
Click to view available options
Size:
Small
Medium
Large
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

The Black Nitrile Glove. Semperit™ Semperguard™ Style Nitrile Gloves are non-sterile, unpowdered, latex-free black nitrile gloves with excellent stretchability, a rolled cuff for easy donning and textured fingers for improved grip.

  • Black nitrile glove with excellent stretchability
  • Particularly suitable for front-end applications in the catering, beauty and body art sector
  • Thanks to dual labeling, also meets the medical requirements of the tattoo and piercing sector
  • Free from phthalates / plasticizers and allergenic latex proteins
  • Rolled cuff enables easy donning
  • Convenient dispenser box holds 100 ambidextrous gloves
  • CE category III protective glove for complex risks, CE class I examination glove
  • Tested and certified according to 1935/2004 - suitable for food contact
Certifications/Compliance Conforms to standards EN 420, EN 374, EN455
Classification CE Category III Protective Glove for Complex Risks, CE Class I Examination Glove 
Color Black
Disposable Single-use
Exterior Finish Fingertip Textured
Grade Certified according to VO 1935/2004 - suitable for contact with food
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size Large
Sterility Non-sterile
Thickness (Metric) 0.14 mm
For Use With (Application) Especially suitable for applications which are visible to the public
Glove Cuff Style Beaded/Rolled
Model Style Nitrile
Packaging Type Dispenser box
Quantity 100/bx
Length (Metric) 240 mm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'