missing translation for 'onlineSavingsMsg'
Learn More

Semperit™ Sempermed™ Synthetic Polyisoprene Syntegra IR Surgical Gloves

Product Code. p-8002746
Click to view available options
Size:
8
8.5
Unit Size:
40 pairs
This item is not returnable. View return policy

Product Code. 12394153

Brand: Semperit™ 3000006141

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Non-latex polyisoprene surgical glove with latex properties is powder-free, latex-free with synthetic internal coating.

  • Made from Synthetic Polyisopren
  • Powder-free
  • Free from natural rubber latex
  • Free from allergenic proteins - this removes the hazard of sensitisation right from the start, especially for
    atopic persons.
  • Quick and easy to put on in every situation due to its elastic material.
  • Lining includes a special net-like structure that reduces friction resistance when donning the gloves
  • Guarantee the easy donning of two pairs of gloves, without any loss of tactile properties.
  • Fully anatomical shape with curved fingers corresponds with the natural, relaxed hand posture
  • Micro-roughened surface offers optimum grip for perfect instrument handling, even in a wet environment

Specifications

Certifications/Compliance EN 1041, EN 980, EN 455/3, ISO 15223, EN 455 Class 1
Color Cream
Disposable Single-use
Latex Free Yes
Material Synthetic Polyisoprene
Powdered Powdered
Size 8.5
Sterility Sterile
Thickness (Metric) 0.19 to 0.24 mm
For Use With (Application) For perfect instrument handling, even in a moist environment, fully anatomical fit guarantee a relaxed hand position for less tiring work
Length (Metric) 285 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.