Learn More
BINDER™ Series FP Avantgarde.Line Drying and Heating Chambers, Gas-tight Option
Product Code. 17804451 Shop All BINDER Products
Click to view available options
For Use With (Equipment):
BINDER Series FP 056 Avantgarde
BINDER Series FP 115 Avantgarde
Unit Size:
Each
Product Code. 17804451
Brand: BINDER™ 80122217
Only null left
Add to basket
Add to basket
The gas tight option model for Series FP Avantgarde.Line Drying and Heating Chambers provides additional measures for the greatest possible impermeability to gas.
  • Accessory for Series FP Avantgarde.Line Drying and Heating Chambers
Specifications
Description Not in conjunction with access port or interior lighting option.
For Use With (Equipment) BINDER Series FP 115 Avantgarde
Type Drying and Heating Chamber
Model Gas-Tight Option
Voltage 120/230 V