missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco™ Serum Bottles
Click to view available options
Capacity (Metric):
2 mL
3 mL
5 mL
10 mL
20 mL
30 mL
50 mL
100 mL
125 mL
Description
- Uniform thin wall construction ensures even freezing and drying
- Ideal containers for use in the FreeZone Stoppering Tray Dryer
- May be connected to valve ports on drying chambers and manifolds
- Supplied in packages of 100
Specifications
Capacity (Metric) | 30 mL |
Color | Clear |
Material | Glass |
Quantity | 100/PK |
Sterility | Non-sterile |
Type | Serum Bottle with 20mm corkage |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'