missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gravis Serum Flasks
Click to view available options
Capacity (Metric):
8 mL
15 mL
24 mL
Unit Size:
Pack of 168
Pack of 224
Pack of 252
Specifications
Specifications
| Capacity (Metric) | 15 mL |
| Diameter (Metric) Outer Neck | 20 mm |
| Material | Glass |
| No. per Pack | 168Pack |
| Quantity | 168Pack |
| Format | Flask |
| Neck Finish | 20mm |
| Product Type | Serum Flask |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction