Learn More
Gravis Serum Flasks
Product Code. p-8009931
Click to view available options
Capacity (Metric):
8 mL
15 mL
24 mL
Unit Size:
Pack of 168
Pack of 224
Pack of 252
Product Code. 11783813
Brand: Gravis F018094672
Only null left
Specifications
Capacity (Metric) 8 mL
Diameter (Metric) Outer Neck 20 mm
Material Glass
No. per Pack 224Pack
Quantity 224Pack
Format Flask
Neck Finish 20mm
Product Type Serum Flask