Learn More
Abeba™ Service 3030 Shoes
A walking sole with cushioning effect, a comfortable footbed and finest upper leather guarantee the highest wearing comfort and a good well being until the end of work.
Brand: Abeba™ 3030/40
Additional Details : Weight : 0.82000kg
Description
- CE EN ISO 20347:2012 O1 FO SRC
- Smooth leather
- Black
- Slip resistance outsole sole
- Integrated footbed
- Lining microfiber, respiration absorbing
- Adjustable instep area
- velcro fastening
Specifications
CE EN ISO 20347:2012 O1 FO SRC | |
Microfiber | |
40 | |
3030 |
Black | |
Leather | |
Slip-resistant |