Learn More
Parker Hannifin™ Service Kit
Product Code. 12567896
Click to view available options
Type:
230 V Solenoid Valve Kit
Unit Size:
Each
Récupel-bijdrage uitgesloten
Product Code. 12567896
Brand: Parker Hannifin™ 606272255
Récupel-bijdrage uitgesloten
36 Month's Service, 230 V Solenoid Valve Kit
Specifications
Includes 1/2 in. BSP solenoid valve (x4), 1/4 in. BSP solenoid valve, coil with 4m cable (x5), M6 fixing bolts (x16) and M6 Washers (x16)
Type 230 V Solenoid Valve Kit