Learn More
Hach Lange™ Set of electrode kit (3 x electrolyte + 3 x membrane)
Product Code. 16468020
Click to view available options
Range:
0.05 to 1000 mg/L NH4-N
Unit Size:
1 set
Product Code. 16468020
Brand: Hach Lange™ LCW868
Only null left
Add to basket
Add to basket
Set of electrode kit (3 x electrolyte + 3 x membrane). For AMTAX sc.
Set of electrode kit (3 x electrolyte + 3 x membrane). For AMTAX sc. Set of electrode kit (3 x electrolyte + 3 x membrane). For AMTAX sc.
Specifications
Instrument: Amtax sc
Parameter: Ammonium
Range: 0.05-1000 mg/L NH4-N
Specifications
Range 0.05 to 1000 mg/L NH4-N
Type Water Testing Kit Supplies and Reagents