Learn More
Hach Lange™ Set of electrode kit (3 x electrolyte + 3 x membrane)
Click to view available options
Range:
0.05 to 1000 mg/L NH4-N
Unit Size:
1 set
Description
Set of electrode kit (3 x electrolyte + 3 x membrane). For AMTAX sc. Set of electrode kit (3 x electrolyte + 3 x membrane). For AMTAX sc.
Specifications
Instrument: Amtax sc
Parameter: Ammonium
Range: 0.05-1000 mg/L NH4-N
Specifications
Specifications
| Range | 0.05 to 1000 mg/L NH4-N |
| Type | Water Testing Kit Supplies and Reagents |