Learn More
Olympus™ Sextuple Revolving Nosepiece
Click to view available options
For Use With (Equipment):
Olympus microscopes
Unit Size:
Each
Description
- Revolving.
- Holds up to 6 objectives.
- Slot for DIC slider.
Specifications
Specifications
| Type | Rotary, Sextuple |
| Device Holds | Objectives, DIC Slider |
| For Use With (Equipment) | Olympus microscopes |