Learn More
Trajan™ 5uL Plunger-in-Needle Syringes
Click to view available options
Volume (Metric):
5 μL
Unit Size:
Each
Description
- High quality solutions for liquid handling
- Accurate and reproducible injections
- 8mm OD barrel with Repeating Adaptor
Specifications
Specifications
| Needle Gauge | 23 G |
| Diameter (Metric) Outer Needle | 0.63 mm |
| Volume (Metric) | 5 μL |
| Needle Point Style | Cone |