Learn More
Trajan™ BP1/BP1 PONA Column
SGE GC column BP1 PONA is an industry standard non polar phase suitable for all routine analysis
Brand:  Trajan™ 054072
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 12985531
  • 1305.00€ / Each
Estimated Shipment: 03-06-2024
to see stock.
Add to basket
Description
Description
  • Classic cross-linked dimethyl polysiloxane technology
  • Low bleed
  • Non-polar
  • Ideal for analyses such as hydrocarbons, aromatics, pesticides and amines
Specifications
Specifications
0.32 mm
Phenols Analysis, Aromatics Analysis, Pesticide Analysis, Phenols, Herbicides Analysis, Amines
1 μm
Non polar
100% Dimethyl Polysiloxane
-60°C to +320/340°C
50 m