Learn More
Trajan™ BPX Dioxin Capillary Columns
SGE BPX Dioxin GC Capillary Columns arespecifically designed for analysis or dioxins and related compounds
Brand:  Trajan™ 054235
Additional Details : Weight : 0.01000kg
Product Code. 15268663
  • 1255.00€ / Each
Estimated Shipment: 12-06-2024
to see stock.
Add to basket
Description
Description
  • High temperature, low bleed column
  • Non-polar
  • Inert
Specifications
Specifications
0.25 mm
0.25 μm
60 m