Learn More
SGE™ ConnecTite™ Liners for Shimadzu™ GC
Product Code. 15279013
Click to view available options
Quantity:
5/Pk.
Product Type:
Connectite Bottom Hole Liner
Connectite Standard Liner
Connectite Top Hole Liner
Unit Size:
Pack of 5
Product Code. 15279013
Brand: SGE™ 092329
Only null left
Add to basket
Add to basket
Designed for use with Shimadzu 2010™ GC
  • Range covers all applications and all popular instrument makes/ models
  • Range covers vaporizing and cool on-column injection techniques
  • Inlet liners ensure complete transfer of sample from syringe to the column without loss
  • 5mm OD, 3.4mm ID; 95mm length
Specifications
Quantity 5/Pk.
Length (Metric) 9.5 cm
Product Type Connectite Standard Liner
Diameter (Metric) Inner 0.34 cm
Diameter (Metric) Outer 0.5 cm
For Use With (Equipment) Shimadzu 2010