Learn More
Trajan™ Methyl Deactivated Fused Silica Capillary Tubing
Product Code. 10781802
Click to view available options
Diameter (English) Inner:
0.002 in.
0.005 in.
0.006 in.
0.008 in.
0.25 mm
0.012 in.
0.020 in.
0.53 mm
Diameter (English) Outer:
0.007 in.
0.008 in.
0.01 in.
0.014 in.
0.016 in.
0.026 in.
0.03 in.
Overall Length (English):
6.56 ft.
6.6 ft.
16.4 ft.
32.8 ft.
82 ft.
Unit Size:
10m
25m
2m
5m
Each
Product Code. 10781802
Brand: Trajan™ 064052
Only null left
Add to basket
Add to basket
Essential for maintaining a high performance chromatography system.Trajan Scientific and Medical™ Methyl Deactivated Fused Silica Capillary Tubing is chemically inert and thermally stable up to 380°C.
  • Suitable for organic and aqueous solvents
  • Methyl deactivated for use with hydrocarbons (pentane, hexane, heptane, iso-octane, aromatics and mixed solvents - n-paraffins/chlorinated solvents
  • Individually tested performance of each 30m length from which all shorter lengths are prepared
  • Ideal for biotechnology applications
Specifications
Type Capillary Tubing
For Use With (Application) Ideal for Biotechnology Applications, and Organic and Aqueous Solvents
Diameter (English) Inner 0.012 in.
Diameter (English) Outer 0.016 in.
Diameter (Metric) Inner 0.32 mm
Diameter (Metric) Outer 0.43 mm
Overall Length (English) 16.4 ft.
Overall Length (Metric) 5 m
Material Fused Silica
Max. Operating Temperature (English) 716°F
Max. Operating Temperature (Metric) 380°C