Learn More
SGE™ eVol™ NMR Starter Kit
Product Code. 12907586
Click to view available options
:
eVol NMR starter kit
Product Code. 12907586
Brand: SGE™ 2910100
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
User-calibrated analytical syringe improves accuracy and reproducibility over standard laboratory tools
  • The programmable digital drive means liquid handling procedures are user-independent, allowing more efficient workflow scheduling and a reduction in the re-analysis of incorrectly processed or false positive samples
  • The gravimetric calibration procedure is simple, intuitive and can be performed at appropriate intervals
  • Kit includes unit, charger, stand, 3 syringes: 5μL with 11.5cm needle, 50μL with 11.5cm and 18cm needles, and 500μL with 11.5cm and 18cm needles
Specifications
For Use With (Application) For user-calibrated analytical syringe improves accuracy and reproducibility over standard laboratory tools
Includes Stand, universal charger, comprehensive, instruction manual, disc with manual in multiple languages, 3 syringes, 5μL syringe with 115mm needle, 50μL syringe with 115mm and 180mm needles, 500μL syringe
Product Type Electronic Syringe Starter Kit