Learn More
Trajan™ eVol™ XCHANGE Syringes
XCHANGE™ analytical syringes can be easily and quickly changed allowing them to be dedicated to individual liquids, and methods, to prevent possible cross-contamination of reagents.
Brand:  Trajan™ 2910329
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 12947596
  • 259.00€ / Pack of 3
Estimated Shipment: 24-05-2024
to see stock.
Description
Description
  • Liquid handling user independent device
  • Reduction in need for re-analysis
  • Syringes easily and quickly changed
  • Replacement parts available
Specifications
Specifications
25 G
1.9 in.
eVol™ Syringe
100 μL
Bevel
Liquid handling procedure
50 mm
3/Pk.
50 mm