missing translation for 'onlineSavingsMsg'
Learn More

SGE™ GC Septa: TCS

Designed specifically for use in Agilent Technologies™ (HP) GC instruments

Brand:  SGE™ 041854

Additional Details : Weight : 0.50000kg

Product Code. 10055570

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.


Description

Description

TCS Septa

  • PTFE-coated triple-layer silicone septa for medium to high temperature applications
  • Good durability, very good resealing
  • Excellent solvent resistance; very good tear resistance
  • Maximum temperature: 280°C (536°F)

Specifications

Specifications

280°C
50/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions