Learn More
Trajan™ Fixed Needle Syringe for Beckman/Altex, Rheodyne, SSI, Valco Valves
missing translation for 'orderingAttributeHoverText'
Volume (Metric):
25 μL
50 μL
250 μL
500 μL
missing translation for 'unitSize'
Each
Description
- High quality solutions for liquid handling
- Accurate and reproducible injections
- Specifically designed for Beckman/Altex, Rheodyne, Valco, SSI valves
Specifications
Specifications
| Material | Borosilicate Glass (Barrel) |
| Needle Gauge | 22 G |
| Diameter (Metric) Inner Needle | 0.37 mm |
| Diameter (Metric) Outer | 0.71 mm |
| Diameter (Metric) Outer Needle | 0.028 mm |
| Product Type | Manual Syringe |
| Volume (Metric) | 25 μL |
| Length (Metric) Needle | 51 mm |
| Needle Point Style | LC |
| For Use With (Equipment) | Beckman/Altex, Rheodyne, SSI, Valco Valves |