Learn More
Trajan™ Replacement Plungers for eVol and eVol NMR Syringes
Product Code. 12967596
Click to view available options
For Use With (Equipment):
eVol NMR Syringe (5μL)
eVol NMR Syringe (50μL)
eVol NMR Syringe (500μL)
Unit Size:
Each
Product Code. 12967596
Brand: Trajan™ 2910380
SGE syringe replacement plungers for gas tight syringes
  • Replacement plungers for gas tight syringes
  • Designed to match the specific syringe barrel to maintain a gas tight seal
Specifications
Product Type Replacement Plunger
For Use With (Equipment) eVol NMR Syringe (5μL)