Learn More
SGE™ SilFlow™ GC 3 Port Splitter Kits
missing translation for 'orderingAttributeHoverText'
Diameter (Metric) Inner:
0.25/0.32 mm
0.53 mm
Port B and C 0.25/0.32 mm
missing translation for 'unitSize'
Each
Description
- Kit includes a SilFlow microchannel device (MCD), SilTite FingerTite tool, mounting bracket, plus appropriate ferrules, nuts and blanking ferrules for set up
- 1.1mm OD tubing not included
Specifications
Specifications
| Type | Port Splitter |
| Diameter (Metric) Inner | 0.53 mm |
| Includes | SilFlow microchannel device (MCD), SilTite FingerTite tool, Mounting bracket, Appropriately sized ferrules, nuts and blanking ferrules for set up |
| For Use With | SGE Silflow Microchannel Device |