Learn More
SGE™ SilTite™ Ferrule Starter Kits Replacement SilTite Nuts and Base Seals
Specially designed ferrule and nut system developed specifically for fused silica GC columns and GC/MS systems
Brand:  SGE™ 073401
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 12700750
  • 530.00€ / Each
Estimated Shipment: 04-06-2024
to see stock.
Add to basket
Description
Description
For use with SGE SilTite Ferrule Starter Kits
SilTite Nuts
  • Non-Swagelok™ nuts with two notches on outer surface
  • Inner taper of the nut corresponds to angle of ferrule, ensuring an even closure of the ferrule around the whole column to eliminate leaks
  • Ferrules are easily removed from nut while still attached to capillary column
  • Reusable
Specifications
Specifications
SilTite Metal Ferrule
Split/Splitless Injector
Inlet Gold Base Seal
Compatible with Agilent Split /Splitless Injector
Metal
10/Pk.