Learn More
Cole-Parmer™ SI-200 Series Stuart™ Shaking Incubator
Product Code. 15847894
Change view
Click to view available options
Holds:
1 L
Large Capacities - 115 L
Medium Capacities - 51 L
Includes:
IQ/OQ
USB Cable
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Includes unitSize
15847894 Large Capacities - 115 L IQ/OQ Each
10257954 1 L USB Cable Each
15827894 Medium Capacities - 51 L IQ/OQ Each
12842163 Large Capacities - 115 L USB Cable Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Regulatory: This product is under eco-contribution tax.
Product Code. 15847894 Supplier Cole-Parmer™ Supplier No. 7952018
Only null left
Add to basket
Add to basket
Regulatory: This product is under eco-contribution tax.
Shaker and incubator combination makes this SI-200 Series Stuart™ Shaking Incubator ideal for cell culture procedures.
  • Microprocessor controlled shaking speed ensures accuracy.
  • Forced air circulation ensures uniform temperature inside the chamber.
  • Easily set and view temperature and speed on the digital display.
  • Unique retractable platform offers easy loading and unloading.
  • These shaking incubators provide an orbital shaking motion to ensure even mixing while promoting bacterial growth.
  • All models feature a timer which can be set from 1 second to 9 days.
  • Once the timer has counted down, an alarm will sound and the shaking action will stop while the incubator will continue to run.
  • The retractable platform allows easy access to the samples at the back of the incubator yet securely locks in place during operation.
  • A versatile clamping system holds flasks securely in place (up to 1 L flasks for 51 L models and up to 2 L flasks for 115 L models).
  • Built-in USB connection and dedicated software allow long-term monitoring of the incubator temperature via a PC.
  • Order models 79520-02 or 79520-18 with IQ/OQ (Installation Qualification/Operational Qualification) documentation to help you comply with regulatory agency requirements.
Specifications
Orbit 16 mm
Speed Range 30 to 300 rpm
Electrical Requirements 230 V
Temperature Range (Metric) 60°C (Max.)
Holds Large Capacities - 115 L
Temperature Range (English) 140°F max
Frequency 50 Hz
Dimensions (LxW) Platform 52.6 x 39 cm
Dimensions (L x W x H) 46.5 x 62.3 x 39.5 cm
Height (English) 15.6 in.
Height (Metric) 39.5 cm
Length (English) 18.3 in.
Length (Metric) 46.5 cm
Width (English) 24.5 in.
Width (Metric) 62.3 cm
Model SI600/IQOQ
Display Digital
For Use With (Application) Cell Culture Applications
Includes IQ/OQ
Timer Range 1 sec. to 9 days