Learn More
Epredia™ Shandon™ Biopsy Processing/Embedding Cassettes II, White
Product Code. 12710928 Shop All Epredia Products
Click to view available options
Color:
Lilac
White
Unit Size:
Each
Pack of 500
Product Code. 12710928
Brand: Epredia™ 1001105
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Hold biopsy specimens during processing and embedding with the special design of Epredia™ Shandon™ Biopsy Processing/Embedding Cassettes II.
  • Acetal polymer construction
  • Locking lid attached by two hinges
  • Cassette writing face is at a 45° angle
  • Pore style is small square
  • Variety of colors available
Ordering Alerts:
  • Supplied in boxes of 500 cassettes with attached lids
TRUSTED_SUSTAINABILITY
Specifications
Pore Style Small Square Pore
Writing Face Angle 45°
Color Lilac
Material Acetal Polymer
Product Type Biopsy Cassette