Learn More
Epredia™ Shandon™ ColorFrost™ Plus Slides
Epredia™ Shandon ColorFrost Plus Slides are positively charged for improved cellular adhesion.
Brand: Epredia™ 9991001
Additional Details : Weight : 0.50000kg
Description
- Electrostatically attract tissue sections and cytology preparations, binding them to the slide
- Eliminate the need for special adhesives or protein coatings
- No background staining
- Available in a variety of colors
Specifications
Microscope Slide | |
Yes | |
100/Cs. |
Glass | |
Blue |