Learn More
Epredia™ Shandon™ Colormark™ Plus Slides
Improve cellular adhesion with positively charged Epredia™ Shandon™ ColorMark™ Plus Slides.
107.00€ - 860.00€
Product Code | Brand | Color | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Quantity | Price | Quantity & Availability | ||||
15394592
|
Epredia™
B9992010 |
White | 144/Pk. |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||||
12996139
|
Epredia™
B9992010AQCS |
Aqua | 1440/Cs. |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||||
12906149
|
Epredia™
B9992010BLCS |
Blue | 1440/Cs. |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||||
12916149
|
Epredia™
B9992010BOCS |
Burnt Orange | 1440/Cs. |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||||
12926149
|
Epredia™
B9992010GLCS |
Gold | 1440/Cs. |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||||
12936149
|
Epredia™
B9992010GRCS |
Green | 1440/Cs. |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||||
12946149
|
Epredia™
B9992010LVCS |
Lavender | 1440/Cs. |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||||
12946219
|
Epredia™
B9992010CS |
Pink | 144/Pk. |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||||
12956149
|
Epredia™
B9992010PKCS |
Pink | 1440/Cs. |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||||
12966149
|
Epredia™
B9992010RDCS |
Red | 1440/Cs. |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||||
12976149
|
Epredia™
B9992010TNCS |
Tan | 1440/Cs. |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||||
12986149
|
Epredia™
B9992010YWCS |
Yellow | 1440/Cs. |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||||
Description
Available in a variety of colors.
Specifications
Glass | |
Microscope Slide |