Learn More
Thermo Scientific™ Shandon™ Consul No-Stick Coverslips
Optimize automated coverslipping with these coverslips, designed to not stick together and ideal for use on the Thermo Scientific Shandon Consul Automated Coverslipper.
Marke: Thermo Scientific™ 7411281
Weitere Details : Weight : 1.00000kg
Beschreibung
Optimize automated coverslipping with Thermo Scientific™ Shandon™ Consul No-Stick Coverslips, designed to not stick together and ideal for use on the Thermo Scientific Shandon Consul Automated Coverslipper.
- 10 x 100 pieces per pack