Learn More
Epredia™ Cryochrome™ Embedding Resin
Product Code. 12580017 Shop All Epredia Products
Click to view available options
Product Type:
Cryochrome Blue
Cryochrome Green
Cryochrome Pink
Cryochrome Sampler Colors
Cryochrome Yellow
Unit Size:
120mL
Each
Pack of 4
Product Code. 12580017
Brand: Epredia™ 9990429
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Support tissue for frozen sectioning with Epredia™ Cryochrome™ Embedding Resin.
  • Color codes frozen sections for easy identification and tracking
  • Four colors available: pink, blue, yellow, green
  • Formulated with physical characteristics identical to Cryomatrix™ embedding resin
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cryochrome Pink
Quantity 120mL