Learn More
Epredia™ Shandon™ Embedding Cassettes II in Tube Packs, aqua
Product Code. 12903755 Shop All Thermo Scientific Products
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
Unit Size:
Pack of 750
Product Code. 12903755
Brand: Epredia™ B1000729AQ
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Hold tissue specimens during processing and embedding with Epredia™ Shandon™ Embedding Cassettes II in Tube Packs.
  • Acetal polymer cassettes with writing face at 45° angle
  • Slotted pore style
  • Locking lid attached by two hinges
Ordering Information:
  • Tube packs are supplied in 10 tubes of 75 cassettes each
TRUSTED_SUSTAINABILITY
Specifications
Pore Style Slotted Pore
Writing Face Angle 45°
Color Aqua
Material Acetal Polymer
Product Type Biopsy Cassette