Learn More
Epredia™ Shandon™ Frozen Section Staining Kit, Glass Beaker
Product Code. 12677716 Shop All Epredia Products
Click to view available options
Type:
Glass Beaker
Unit Size:
Pack of 12
Product Code. 12677716
Brand: Epredia™ 146
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Glass Beaker for use with the Epredia™ Shandon™ Frozen Section Staining Kit.
  • Glass beakers have a capacity of 200mL (6.75 oz.) and are 10cm (4 in.) tall
TRUSTED_SUSTAINABILITY
Specifications
Material Glass
Type Glass Beaker