Learn More
Epredia™ Shandon™ Octospot™ Accessories
Shop All Epredia Products
Click to view available options
Type:
8-well Strips
8-well Strips, Loose
EZ-Write Slides, 8 spots
Holding Block
Solvent-Resistant Slides, 8 spots
Thick Filter Cards
Unit Size:
Each
Pack of 100
Pack of 5
Pack of 60
Specifications
Specifications
| Type | Solvent-Resistant Slides, 8 spots |
| For Use With (Equipment) | Shandon™ Octospot™ Starter Packs |