Learn More
Epredia™ Shandon™ Vertical Staining Jar, Vertical staining jar, Holds 16 slides back-to-back
Shop All Epredia ProductsDescription
Features:
- Heavy, clear glass construction
- Internal grooves
- Wide-mouthed for easy slide handling
- 1.85 x 1.85in. (47 x 47mm) square, flanged top
- Flanged, clear glass cover
Specifications
Specifications
| Closure Included | Closure Included |
| Closure Material | Glass |
| Bottom Shape | Flat |
| Color | Clear |
| Material | Glass |
| Type | Staining Jar |
| Includes | Cover |
| Slide Count | 16 |
| Length (English) | 1.85 in. |
| Length (Metric) | 47 mm |
| Show More |