missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ SHARPFLEX Latex Gloves

Product Code. 15836745
Click to view available options
Size:
7
8
9
10
Unit Size:
1 pair
This item is not returnable. View return policy

Product Code. 15836745

Brand: Honeywell™ 223252508

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Honeywell™ SHARPFLEX Latex Gloves for handling of heavy parts with high cut risks in humid and slippery environments.

  • Knitted glove in composite and para-aramid yarn
  • The knitting technique provides the glove with excellent fit and high dexterity
  • The composite yarn is reliable and safe protection
  • The para-aramid yarn assure a resistance to the contact heat and cut risks
  • Plated polyamide inside
  • Offer a greater comfort and limit the contact of the skin with composite fiber
  • Blue crinkled latex coating on palm
  • Latex coating provides better grip and a light thermal protection (up to 250°C) resistance
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN 420-03 EEC Basic standards, EN 388-03 Mechanical hazards, EN407-04 : Thermal risks
Description Para-aramid and composite, 13 gauge
Color Yellow, Green, Blue
Material Latex
Size 8
Length (Metric) 25 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.